2,4-Dihydroxybenzoic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: beta-Resorcylic acid

121295-100G 121295-2.5KG 121295-500G
BT121295-100GEA 25 USD
BT121295-100G BT121295-2.5KG BT121295-500G
2,4-Dihydroxybenzoic acid ≥97%
2,4-Dihydroxybenzoic acid

CAS: 89-86-1; EC No: 201-946-9; MDL No: MFCD00002451; RTECS: VH3708050 Crystalline/Powder; Molecular Formula: C7H6O4; MW: 154.12 Melting Point: 208-211° (decomposes)


  • RTECS: VH3708050
  • Crystalline/Powder

Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 208…211 °C (dec.)
MDL Number: MFCD00002451
CAS Number: 89-86-1
EINECS: 201-946-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR