Calcium formate ≥98%

Supplier: BeanTown Chemical

Synonyms: Formic acid calcium salt

121270-100G 121270-500G
BT121270-100GEA 47.78 USD
BT121270-100G BT121270-500G
Calcium formate ≥98%
Calcium formate

CAS: 544-17-2; EC No: 208-863-7; MDL No: MFCD00036108; RTECS: LQ5600000 Powder; Molecular Formula: C2H2CaO4; MW: 130.11


  • RTECS: LQ5600000
  • Powder

Formula: C₂H₂CaO₄
MDL Number: MFCD00036108
CAS Number: 544-17-2
EINECS: 208-863-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR