8-Aminocaprylic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
ω-Aminooctoic acid, 8-Aminooctanoic acid, ω-Aminocaprylic acid, ω-Aminooctanoic acid, 8-Aminooctoic acid
121225-1G
121225-5G
BT121225-1GEA
50
USD
BT121225-1G
BT121225-5G
8-Aminocaprylic acid ≥99%
8-Aminocaprylic acid
CAS: 1002-57-9; EC No: 213-687-9; MDL No: MFCD00008245
Crystalline/Powder; Linear Formula: NH2(CH2)7CO2H; Molecular Formula: C8H17NO2; MW: 159.23
Melting Point: 194° (decomposes)
- Crystalline/Powder
Formula:
C₈H₁₇NO₂ MW: 159.23 g/mol Melting Pt: 194 °C (dec.) |
MDL Number:
MFCD00008245 CAS Number: 1002-57-9 EINECS: 213-687-9 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...