N,N'-Diphenylbenzidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 4,4'-Bis(phenylamino)biphenyl, N4,N4'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine

121215-25G 121215-5G
BT121215-25GEA 206 USD
BT121215-25G BT121215-5G
N,N'-Diphenylbenzidine ≥98%
N,N'-Diphenylbenzidine

CAS: 531-91-9; EC No: 208-521-7; MDL No: MFCD00003016 Powder; Linear Formula: (C6H5NH)C6H4C6H4(NHC6H5); Molecular Formula: C24H20N2; MW: 336.44 Melting Point: 246-248° Light Sensitive


  • Light Sensitive
  • Powder

Formula: C₂₄H₂₀N₂
MW: 336.44 g/mol
Melting Pt: 246…248 °C
MDL Number: MFCD00003016
CAS Number: 531-91-9
EINECS: 208-521-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR