Phenazine ≥99%
Supplier: BeanTown Chemical
121195-100G
121195-25G
121195-5G
BT121195-100GEA
435
USD
BT121195-100G
BT121195-25G
BT121195-5G
Phenazine ≥99%
Phenazine
CAS: 92-82-0; EC No: 202-193-9; MDL No: MFCD00005023; RTECS: SG1360000
Crystalline/Powder; Molecular Formula: C12H8N2; MW: 180.21
Melting Point: 172-176°; Boiling Point: 360°
- RTECS: SG1360000
- Crystalline/Powder
Formula:
C₁₂H₈N₂ MW: 180.21 g/mol Boiling Pt: 360 °C Melting Pt: 172…176 °C |
MDL Number:
MFCD00005023 CAS Number: 92-82-0 EINECS: 202-193-9 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...