Tris[3,5-bis(trifluoromethyl)phenyl]phosphine ≥95%

Supplier: BeanTown Chemical

Synonyms: Tris(3,5-bis(trifluoromethyl)phenyl)phosphine

121190-1G 121190-250MG
BT121190-1GEA 155 USD
BT121190-1G BT121190-250MG
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine ≥95%
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine

CAS: 175136-62-6; MDL No: MFCD00042428 Powder; Linear Formula: [(CF3)2C6H3]3P; Molecular Formula: C24H9F18P; MW: 670.28 Melting Point: 98-102° Air Sensitive


  • Air Sensitive
  • Powder

Formula: C₂₄H₉F₁₈P
MW: 670.28 g/mol
Melting Pt: 98…102 °C
MDL Number: MFCD00042428
CAS Number: 175136-62-6

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR