Aluminium lanthanum (III) oxide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Lanthanum(III) aluminum oxide, Lanthanum Aluminium oxide, Aluminium lanthanum trioxide

121180-25G 121180-5G
BT121180-25GEA 164 USD
BT121180-25G BT121180-5G
Aluminium lanthanum (III) oxide ≥99.9%
Aluminium lanthanum (III) oxide

CAS: 12003-65-5; EC No: 234-433-3; MDL No: MFCD00144550 Powder; Linear Formula: LaAlO3 ; MW: 213.88


  • Powder

Formula: AlLaO₃
MW: 213.89
MDL Number: MFCD00144550
CAS Number: 12003-65-5
EINECS: 234-433-3

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR