5-Bromo-2-chloro-3-fluoropyridine ≥98%

Supplier: BeanTown Chemical

121165-25G 121165-5G
BT121165-25GEA 336 USD
BT121165-25G BT121165-5G
5-Bromo-2-chloro-3-fluoropyridine ≥98%
5-Bromo-2-chloro-3-fluoropyridine

CAS: 831203-13-5; MDL No: MFCD03411574 Solid; Molecular Formula: C5H2BrClFN ; MW: 210.43


  • Solid

Formula: C₅H₂BrClFN
MW: 210.43 g/mol
MDL Number: MFCD03411574
CAS Number: 831203-13-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR