Agar Powder

Supplier: BeanTown Chemical

121155-100G 121155-2.5KG 121155-500G
BT121155-100GEA 35.62 USD
BT121155-100G BT121155-2.5KG BT121155-500G
Agar Powder
Media for Microbiology Dehydrated Media for Microbiology

Molecular formula: (C₁₂H₁₈O₉)n
Merck number: 14,184
EC number: 232-658-1


Información de suministro: Non-hazardous for shipping.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR