Polyvinylpyrrolidone cross-linked

Supplier: BeanTown Chemical

Synonyms: Polyvinylpolypyrrolidone, N-Vinyl-2-Pyrrolidone

121135-100G 121135-500G
BT121135-100GEA 57.54 USD
BT121135-100G BT121135-500G
Polyvinylpyrrolidone cross-linked
Polyvinylpyrrolidone cross-linked

CAS: 25249-54-1; MDL No: MFCD00149016 Powder; Molecular Formula: (C6H9NO)n Melting Point: 300°


  • Powder

Formula: C₆H₉NO
MW: 111.144 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00149016
CAS Number: 25249-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR