Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, Ethyl 4-chloro-2-(methylsulphanyl)pyrimidine-5-carboxylate, Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate

121130-100G 121130-25G 121130-5G
BT121130-100GEA 837 USD
BT121130-100G BT121130-25G BT121130-5G
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate ≥98%
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate

CAS: 5909-24-0; EC No: 227-619-0; MDL No: MFCD00006085 Powder; Molecular Formula: C8H9ClN2O2S; MW: 232.69 Melting Point: 60-63°


  • Powder

Formula: C₈H₉ClN₂O₂S
MW: 232.69 g/mol
Melting Pt: 60…63 °C
MDL Number: MFCD00006085
CAS Number: 5909-24-0
EINECS: 227-619-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR