6-Bromoindole ≥98%
Supplier: BeanTown Chemical
Synonyms:
6-Bromo-1H-indole
121115-1G
121115-250MG
BT121115-1GEA
211
USD
BT121115-1G
BT121115-250MG
6-Bromoindole ≥98%
6-Bromoindole
CAS: 52415-29-9; MDL No: MFCD00238550
Solid; Molecular Formula: C8H6BrN; MW: 196.04
Melting Point: 92-96°
- Solid
Formula:
C₈H₆BrN MW: 196.05 g/mol Melting Pt: 92…96 °C |
MDL Number:
MFCD00238550 CAS Number: 52415-29-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...