Iron(II) gluconate hydrate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Iron(II) Gluconate Hydrate, Iron(II) D-gluconate dihydrate, Gluconic Acid Iron(II) Salt
121105-2KG
121105-500G
BT121105-2KGEA
145
USD
BT121105-2KG
BT121105-500G
Iron(II) gluconate hydrate ≥97%
Iron(II) gluconate hydrate
CAS: 22830-45-1; EC No: 206-076-3; MDL No: MFCD00150872; RTECS: LZ5180000
Granular; Linear Formula: Fe[HOCH2(CHOH)4CO2]2·xH2O; Molecular Formula: C12H22FeO14; MW: 446.19 (anhydrous)
Melting Point: 188° (decomposes)
- RTECS: LZ5180000
- Granular
Formula:
C₁₂H₂₄FeO₁₅ MW: 464.16 g/mol Melting Pt: 188 °C (dec.) |
MDL Number:
MFCD00150872 CAS Number: 22830-45-1 EINECS: 206-076-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...