3-Hydroxypicolinic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Hydroxypyridine-2-carboxylic acid

121070-1G 121070-5G
BT121070-1GEA 35 USD
BT121070-1G BT121070-5G
3-Hydroxypicolinic acid ≥98%
3-Hydroxypicolinic acid

CAS: 874-24-8; EC No: 212-859-0; MDL No: MFCD00006294 Powder; Molecular Formula: C6H5NO3; MW: 139.11 Melting Point: 213-218°


  • Powder

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 213…218 °C
MDL Number: MFCD00006294
CAS Number: 874-24-8
EINECS: 212-859-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR