Pentafluorophenylboronic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,3,4,5,6-Pentafluorobenzeneboronic acid, Dihydroxy(pentafluorophenyl)borane, (Pentafluorophenyl)metaboric acid, Pentafluorobenzeneboronic acid
121055-100G
121055-25G
121055-5G
BT121055-100GEA
1134
USD
BT121055-100G
BT121055-25G
BT121055-5G
Pentafluorophenylboronic acid ≥97%
Pentafluorophenylboronic acid
CAS: 1582-24-7; EC No: 000-000-0; MDL No: MFCD01074663
Solid; Molecular Formula: C6H2BF5O2; MW: 211.88
Moisture Sensitive
- Moisture Sensitive
- Solid
Formula:
C₆H₂BF₅O₂ MW: 211.88 g/mol |
MDL Number:
MFCD01074663 CAS Number: 1582-24-7 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...