Iron(III) citrate hydrate

Supplier: BeanTown Chemical

Synonyms: Ferric citrate

121050-1KG 121050-250G 121050-50G
BT121050-1KGEA 156.89 USD
BT121050-1KG BT121050-250G BT121050-50G
Iron(III) citrate hydrate
Iron(III) citrate hydrate

CAS: 334024-15-6; EC No: 222-536-6; MDL No: MFCD00149617 Solid; Molecular Formula: C6H5FeO7·xH2O; MW: 244.94 (anhydrous) Light Sensitive

Formula: C₆H₅FeO₇·xH₂O
MW: 244.948 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149615
CAS Number: 334024-15-6
EINECS: 222-536-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR