Diyttrium trioxide ≥99.99% (trace rare earth metals basis)
Supplier: BeanTown Chemical
Synonyms:
Yttrium oxide, Yttrium (III) oxide
121045-100G
121045-25G
121045-500G
BT121045-100GEA
110
USD
BT121045-100G
BT121045-25G
BT121045-500G
Diyttrium trioxide ≥99.99% (trace rare earth metals basis)
Diyttrium trioxide
CAS: 1314-36-9; EC No: 215-233-5; MDL No: MFCD00011473; RTECS: ZG3850000
- RTECS: ZG3850000
- Air Sensitive
- <10 Micron Powder
Formula:
Y₂O₃ MW: 225.81 g/mol Boiling Pt: 4300 °C Melting Pt: 2410 °C Density: 5.01 g/mL |
MDL Number:
MFCD00011473 CAS Number: 1314-36-9 EINECS: 215-233-5 |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...