2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-[N,N-Bis(Trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)triflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonyl)imide, Comins Triflating Reagent, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonimide)
121005-1G
121005-5G
BT121005-1GEA
44
USD
BT121005-1G
BT121005-5G
2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine ≥99%
2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine
CAS: 145100-51-2; EC No: 000-000-0; MDL No: MFCD00191833
Crystalline; Molecular Formula: C7H3ClF6N2O4S2; MW: 392.68
Melting Point: 45-47°
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
C₇H₃ClF₆N₂O₄S₂ MW: 392.67 g/mol Melting Pt: 45…47 °C |
MDL Number:
MFCD00191833 CAS Number: 145100-51-2 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...