Immun-Blot® Low Fluorescence PVDF Membrane, Bio-Rad
Supplier: Bio-Rad
1620260 1620261 1620262 1620263 1620264
102650-428EA 250.77 USD
102650-428 102650-430 102650-432 102650-434 102650-436
Immun-Blot® Low Fluorescence PVDF Membrane, Bio-Rad
Transfer Membranes
Immun-Blot® Low Fluorescence PVDF Membrane has the same properties as standard PVDF membranes coupled with minimal autofluorescence.
  • Low autofluorescence allows long exposure times and maximizes the sensitivity of fluorescent detection
  • High protein retention and broad dynamic range
  • Compatible with multiplex fluorescent protocols
  • Compatible with chemiluminescent and colorimetric detection methods
Immun-Blot® Low Fluorescence PVDF Membrane is available in precut membrane and filter paper sets, and rolls.