Cap Strips for PCR Tubes and Plates, Bio-Rad
Supplier: Bio-Rad
TCS0801 TCS1201 TCS0803
101647-292EA 83.08 USD
101647-292 101647-296 101647-294
Cap Strips for PCR Tubes and Plates, Bio-Rad
Closures Test/Sample Tube Closures
Cap strips provide extremely tight sealing of all Bio-Rad 0.2 ml PCR tubes and plates during thermal cycling and cold storage
Flat cap strips feature ultraclear upper surfaces that are ideal for light-based assays such as real-time PCR. Average light transmittance is 1.7-fold higher than with standard clarity domed cap strips.
Caps are a good option to save plates if fewer wells are needed during PCR. Use of a capping tool is recommended for seating of caps on tubes or plates.
Flat caps are available in strips of 8, and domed caps are available in strips of 8 or 12. Caps are process-sampled and tested and are free of DNase, RNase, and human DNA.