Diphenylboric acid-2-aminoethyl ester (2-APB), Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
2-Aminoethyl diphenylborinate, 2-((Diphenylboryl)oxy)ethanamine
M-Clarity™ quality level = MQ200
8.41636.0005
8.41636.8500
EM8.41636.0005CS
83797
USD
EM8.41636.0005
EM8.41636.8500
Diphenylboric acid-2-aminoethyl ester (2-APB), Sigma-Aldrich®
Diphenylboric acid-2-aminoethyl ester (2-APB)
Formula:
C₁₄H₁₆BNO |
MDL Number:
MFCD00014823 CAS Number: 524-95-8 |
Specification Test Results
CAS | 524-95-8 |
Synonyms | Diphenylboric acid-2-aminoethyl ester |
Hill Formula | C₁₄H₁₆BNO |
Molar Mass | 225.09 g/mol |
Melting Point | 189 - 194 °C |
Storage | Store below +15°C. |
Assay (HClO₄) | ≥ 98,0 %(m) |
Identity (IR) | conforms |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...