Adipohydrazide, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Adipyl hydrazide

M-Clarity™ quality level = MQ200

8.41689.0050 8.41689.0010
EM8.41689.0050CS 3378 USD
EM8.41689.0050 EM8.41689.0010
Adipohydrazide, Sigma-Aldrich®
Adipohydrazide
Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Boiling Pt: 446
MDL Number: MFCD00007614
CAS Number: 1071-93-8

Order Now

Specification Test Results

CAS 1071-93-8
Hill Formula C₆H₁₄N₄O₂
Molar Mass 174.2 g/mol
Flash point less than 150 °C
Ignition temperature 360 °C
Melting Point 178 - 182 °C
Assay (HClO₄) ≥ 97,0 %(m)
Melting range (lower value) ≥ 178 °C
Melting range (upper value) ≤ 182 °C
Identity (IR) conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR