Premixed Transfer Buffers, Western Blotting, Bio-Rad
Supplier: BIO-RAD LABORATORIES MS
1610734 1610771 1610778
101643-364EA 60 USD
101643-364 101643-402 101643-412
Premixed Transfer Buffers, Western Blotting, Bio-Rad
Buffers Electrophoresis Buffers Transfer Buffers
High-quality, ready-to-run transfer buffers in 1 L bottles and economical 5 L cubes.
Three premixed transfer buffers are available: 10x Tris/glycine, 10x Tris/CAPS, and 20x SSC. Generally, 10x Tris/glycine is used for standard wet/tank blotting, and 10x Tris/CAPS for semi-dry blotting.