Molecular Rulers, DNA Standards, Bio-Rad
Supplier: BIO-RAD LABORATORIES MS
1708200 1708202 1708353 1708355 1708352 1708204 1708206 1708205 1708201 1708203 1708354 1708351
101645-320EA 367.69 USD
101645-320 101645-324 101645-358 101645-362 101645-356 101645-328 101645-332 101645-330 101645-322 101645-326 101645-360 101645-354
Molecular Rulers, DNA Standards, Bio-Rad
Electrophoresis Reagents Molecular Weight Markers
Bio-Rad provides three types of molecular rulers: Standard, EZ Load™, and AmpliSize® Rulers
Standard Molecular Rulers are precisely sized DNA, ready for dilution. EZ Load™ Molecular Rulers are ready-to-load. They include a vial of 5X sample buffer for preparing your own samples. AmpliSize® Molecular Rulers contain ten bands (at 50, 100, 200, 300, 400, 500, 700, 1,000, 1,500, and 2,000 bp) blunt-ended DNA with 50% GC content.
To ensure correct sizing of bands in samples, most of these DNA ladders include a visually distinct reference band.