Matrix™ Deepwell Storage Blocks, Thermo Scientific
Supplier: Thermo Fisher Scientific
4222 4325 4211-11 4212-11 4221
101504-424CS 750.46 USD
101504-424 101504-494 101504-418 101504-420 101504-422
Matrix™ Deepwell Storage Blocks, Thermo Scientific
Microplates Deep Well Microplates
Thermo Scientific™ Matrix™ Deepwell Storage Blocks are constructed of polypropylene and are chemically resistant.
The unique chimney well design of the 1 mL block reduces the risk of cross-contamination between wells during pipetting, while the 2 mL block has cut-away sides for easy handling and placement on robotic systems. Meets all standard microplate specifications.