Human Recombinant IL-17E (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-149-100UG 100-149-1MG 100-149-25UG 100-149-5UG 100-149-500UG
10787-356EA 564.48 USD
10787-356 10787-358 10787-360 10787-362 10822-170
Human Recombinant IL-17E (from E. coli)
Proteins and Peptides
Interleukin 17E (IL-17E), also known as IL-25, is a member of the IL-17 family of cytokines. IL-17E binds to the IL-17RB receptor to stimulate the secretion of the proinflammatory interleukin 8 (IL-8) chemokine and to induce the activation of nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB).
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.