Guardian Grid Tape, Transforming Technologies
Supplier: Transforming Technologies
CT2406 CT1805 CT1204 CT4807
14224-682CS 63.5 USD
14224-682 14224-686 14224-680 14224-688
Guardian Grid Tape, Transforming Technologies
Tapes
Our proprietary three-layer tape provides non-sparking surfaces on both the inside and the outside
  • Applications include use ESD symbols for ESD awareness, sealing ESD packaging/containers, for applications requiring EMI shielding, use in a grounded tape dispenser, voltage generated by unrolling will effectively be reduced to zero.
A conductive grid is buried between two static dissipating copolymer substrates that will not shed, crack, chip or rub off. Available on 3" cores.
Certificaciones: Meets or exceeds requirements of ANSI ESD-S20.20.