Ear Punch, Roboz Surgicals
Supplier: Roboz Surgical
65-9902 65-9901 65-9900
100188-948EA 65.08 USD
100188-948 100188-946 100189-750
Ear Punch, Roboz Surgicals
Biopsy Punches
Ear punching is a simple and inexpensive way to identify laboratory animals.
  • No need to stock additional ear tags
  • Small holes mark ears of small lab animals
There is no need to stock additional ear tags or other supplies. These ear punches marks the ears of small laboratory animals by making holes.