Spatulas, Tapered, Micro, #304 Stainless Steel, Non-Magnetic, Chemglass
Supplier: Chemglass
CG-1982-11 CG-1982-12
80071-668EA 9.71 USD
80071-668 80071-670
Spatulas, Tapered, Micro, #304 Stainless Steel, Non-Magnetic, Chemglass
Spatulas
Spatulas are made of non-magnetic stainless steel and feature a mirror finish.
  • Spatula will pass through a 14/10 or 14/20 outer joint
  • One end is rounded, the other is tapered to ¹/₈" (3.3 mm)
Both ends are approx 8 mm (⁵/₁₆") wide × 51 mm (2") in length.