KWIK-STIK™ Plus Microorganisms, Microbiologics
Supplier: MICROBIOLOGICS, INC.
0235X 0255X 0258X 0306X 0317X 0335X 0351X 0353X 0360X 0363X 0366X 0369X 0370X 0371X 0385X 0392X 0419X 0421X 0443X 0483X 0484X 0485X 0486X 0487X 0488X 0500X 0647X 0650X 0671X 0678X 0683X 0684X 0687X 0688X 0689X 0693X 0698X 0699X 0805X 0806X 0813X 0827X 0836X 0852X 0871X 0890X 0902X 0919X 0947X 0998X 01110X 0524X
89501-820EA 453.48 USD
89501-820 89502-846 89503-058 89503-162 89503-184 89503-428 89503-506 89503-636 89503-732 89503-758 89503-826 89503-850 89503-886 89503-910 89503-918 89503-946 89504-026 89504-094 89504-256 89504-276 89504-392 89504-610 89504-632 89504-660 89504-692 89504-712 89504-724 89504-790 89505-444 89505-466 89505-546 89505-580 89505-614 89505-626 89505-658 89505-682 89505-694 89505-732 89505-758 89505-770 89506-328 89506-338 89506-390 89506-466 89506-524 89506-622 89506-728 89506-818 89506-882 89506-952 89507-042 89507-332 10119-782 10127-626
KWIK-STIK™ Plus Microorganisms, Microbiologics
Controls and Calibration Verification Materials
KWIK-STIK Plus microorganisms are designed to meet the needs of laboratories requiring QC microorganisms that are a limited number of passages from the reference culture. Each KWIK-STIK Plus QC microorganism preparation is just two passages from the reference culture, allowing for more subcultures. Conveniently packaged in a KWIK-STIK device featuring a lyophilized QC microorganism pellet, inoculating swab and reservoir of hydrating fluid, KWIK-STIK Plus is ready to use and affordable. For added convenience, a peel-off Certificate of Assay is included for quality control documentation.
KWIK-STIK Plus includes five STIKS of a single strain
Online Certificate of Analysis provides documentation of phenotypic, macroscopic and microscopic test results.
Two passages from reference culture allows more subculturing.
Certificaciones: Traceable to reference culture. Assured quality.
Información para pedidos: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.