2,3,5-Tri-O-benzyl-D-arabinofuranose

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R,4R)-2,3,5-Tris(benzyloxy)-4-hydroxypentanal

J66792-09 J66792-06
AAJ66792-09EA 305 USD
AAJ66792-09 10814-858 10815-832
2,3,5-Tri-O-benzyl-D-arabinofuranose
2,3,5-Tri-O-benzyl-D-arabinofuranose

Powder.


CAS No.: 37776-25-3 Molecular Formula: C26H28O5 Formula Weight: 420.50 Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00080813

Formula: C₂₆H₂₈O₅
MW: 420.505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080813
CAS Number: 37776-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR