2,3,5-Tri-O-benzyl-b-D-ribofuranose 98%, powder

Supplier: Thermo Scientific Chemicals

J66252-06 J66252-03
AAJ66252-04EA 188.14 USD
AAJ66252-04 AAJ66252-06 10814-530 10814-532
2,3,5-Tri-O-benzyl-b-D-ribofuranose 98%, powder
2,3,5-Tri-O-benzyl-b-D-ribofuranose

2,3,5-Tri-O-benzyl-^b-D-ribofuranose, 98%. Powder.


Purity: 98% CAS No.: 89361-52-4 Molecular Formula: C26H28O5 Formula Weight: 420.50 Physical Form: Powder Appearance: White Solubility: Soluble in chloroform Optical Rotation: +43° (c=2 in dioxane) MDL No.: MFCD01864312

Formula: C₂₆H₂₈O₅
MW: 420.505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01864312
CAS Number: 89361-52-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR