3,4-Diaminobenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H66954-03 H66954-06 H66954-14
AAH66954-03EA 38.6 USD
AAH66954-03 AAH66954-06 AAH66954-14
3,4-Diaminobenzonitrile 97%
3,4-Diaminobenzonitrile

3,4-Diaminobenzonitrile, 97%


Purity: 97% CAS No.: 17626-40-3 Molecular Formula: C7H7N3 Formula Weight: 133.15 Physical Form: Solid Appearance: Pale brown Melting Point: 140-148° UN No.: 3439 MDL No.: MFCD00723901

Formula: C₇H₇N₃
Melting Pt: 140…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00723901
CAS Number: 17626-40-3
UN: 3439
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR