Fmoc-val-OSu 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-L-valine N-succinimidyl ester, Fmoc-Val-OSu
H66946-06
H66946-14
AAH66946-06EA
58
USD
AAH66946-06
AAH66946-14
Fmoc-val-OSu 95%
Fmoc-val-OSu
N-Fmoc-L-valine N-succinimidyl ester, 95%
Purity: 95%
CAS No.: 130878-68-1
Molecular Formula: C24H24N2O6
Formula Weight: 436.46
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white to white
MDL No.: MFCD00153370
Formula:
C₂₄H₂₄N₂O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00153370 CAS Number: 130878-68-1 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...