Fmoc-Gly-Gly-OH
Supplier: Thermo Scientific Chemicals
H66923-03
H66923-06
AAH66923-03EA
40.1
USD
AAH66923-03
AAH66923-06
Fmoc-Gly-Gly-OH
Proteins and Peptides
N-Fmoc-L-glycylglycine, 97%
Purity: 97%
CAS No.: 35665-38-4
Molecular Formula: C19H18N2O5
Formula Weight: 354.36
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 2228504
MDL No.: MFCD00190880
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...