Isoxazole-4-boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 4-Isoxazoleboronic acid pinacol ester

H66887-03 H66887-MD
AAH66887-03EA 157 USD
AAH66887-03 AAH66887-MD
Isoxazole-4-boronic acid pinacol ester 97%
Isoxazole-4-boronic acid pinacol ester

Isoxazole-4-boronic acid pinacol ester, 97%


Purity: 97% CAS No.: 928664-98-6 Molecular Formula: C9H14BNO3 Formula Weight: 195.02 Melting Point: 110-115° MDL No.: MFCD06657891

Formula: C₉H₁₄BNO₃
MW: 195.02 g/mol
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD06657891
CAS Number: 928664-98-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR