2-Amino-5-fluorobenzamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Fluoroanthranilamide
H66873-03
H66873-06
AAH66873-03EA
56.5
USD
AAH66873-03
AAH66873-06
AAH66873-MD
2-Amino-5-fluorobenzamide 97%
2-Amino-5-fluorobenzamide
2-Amino-5-fluorobenzamide, 97%
Purity: 97%
CAS No.: 63069-49-8
Molecular Formula: C7H7FN2O
Formula Weight: 154.14
Physical Form: Crystalline
Appearance: Off-white
Melting Point: 143-148°
Beilstein Reference: 2718500
MDL No.: MFCD03428522
Formula:
C₇H₇FN₂O MW: 154.14 g/mol Melting Pt: 143…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD03428522 CAS Number: 63069-49-8 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...