Fmoc-Cys(Me)-OH 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Fmoc-S-methyl-L-cysteine
H66842-06
H66842-14
AAH66842-06EA
73.2
USD
AAH66842-06
AAH66842-14
Fmoc-Cys(Me)-OH 95%
Fmoc-Cys(Me)-OH
N-Fmoc-S-methyl-L-cysteine, 95%
Purity: 95%
CAS No.: 138021-87-1
Molecular Formula: C19H19NO4S
Formula Weight: 357.42
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00237014
Formula:
C₁₉H₁₉NO₄S Storage Temperature: Refrigerator |
MDL Number:
MFCD00237014 CAS Number: 138021-87-1 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...