Boc-D-Tyr-OMe 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-D-tyrosine methyl ester
H66827-06
H66827-14
AAH66827-06EA
52
USD
AAH66827-06
AAH66827-14
AAH66827-22
Boc-D-Tyr-OMe 96%
Boc-D-Tyr-OMe
N-Boc-D-tyrosine methyl ester, 96%
Purity: 96%
CAS No.: 76757-90-9
Molecular Formula: C15H21NO5
Formula Weight: 295.34
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 4704567
MDL No.: MFCD00057824
Formula:
C₁₅H₂₁NO₅ MW: 295.335 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00057824 CAS Number: 76757-90-9 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...