Fmoc-Lys(Ddiv)-OH 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Nα-Fmoc-Nepsilon-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]-L-lysine, Fmoc-Lys(ivDde)-OH

H66820-03 H66820-MD
AAH66820-03EA 215 USD
AAH66820-03 AAH66820-MD
Fmoc-Lys(Ddiv)-OH 95%
Fmoc-Lys(Ddiv)-OH

N^a-Fmoc-N^e-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]-L-lysine, 95%


Purity: 95% CAS No.: 204777-78-6 Molecular Formula: C34H42N2O6 Formula Weight: 574.72 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD01631658

Formula: C₃₄H₄₂N₂O₆
MW: 574.718 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631658
CAS Number: 204777-78-6

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR