2-Amino-5-methoxypyrimidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
2-Amino-5-methoxypyrimidine, 97%
Purity: 97%
CAS No.: 13418-77-4
Molecular Formula: C5H7N3O
Formula Weight: 125.13
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: Off-white
Beilstein Reference: 607358
MDL No.: MFCD00187864
Formula:
C₅H₇N₃O Storage Temperature: Ambient |
MDL Number:
MFCD00187864 CAS Number: 13418-77-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...