N-Acetyl-L-isoleucine 98% (by HPLC)

Supplier: Thermo Scientific Chemicals

Synonyms: Ac-Ile-OH, N-Acetyl-L-Isoleucine, (2S,3S)-2-Acetamido-3-methylpentanoic acid

H66771-06 H66771-14
AAH66771-06EA 74.8 USD
AAH66771-06 AAH66771-14 AAH66771-22
N-Acetyl-L-isoleucine 98% (by HPLC)
N-Acetyl-L-isoleucine

N-Acetyl-L-isoleucine, 98%


Purity: 98% CAS No.: 3077-46-1 Molecular Formula: C8H15NO3 Formula Weight: 173.21 Storage Temperature: -30°C to -10°C Physical Form: Crystalline Appearance: White to off-white MDL No.: MFCD00066072

Formula: C₈H₁₅NO₃
MW: 173.212 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00066072
CAS Number: 3077-46-1
EINECS: 221-364-9

Order Now

Specification Test Results

Assay (HPLC) >97.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR