IR-775 chloride 90% (dye content)

Supplier: Thermo Scientific Chemicals

H66770-03 H66770-MD
AAH66770-03EA 251 USD
AAH66770-03 AAH66770-MD
IR-775 chloride 90% (dye content)
IR-775 chloride

Purity: 90% CAS No.: 199444-11-6 Molecular Formula: C32H36Cl2N2 Formula Weight: 519.55 Physical Form: Solid Appearance: Pale green to dark green Melting Point: 205-214° MDL No.: MFCD03427067

Formula: C₃₂H₃₆Cl₂N₂
Melting Pt: 205…214 °C
Storage Temperature: Ambient
MDL Number: MFCD03427067
CAS Number: 199444-11-6

Order Now

Specification Test Results

Assay Minimum 90


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR