Fmoc-Bpa-OH 95%
Supplier: Thermo Scientific Chemicals
N-Fmoc-4-benzoyl-L-phenylalanine, 95%
Purity: 95%
CAS No.: 117666-96-3
Molecular Formula: C31H25NO5
Formula Weight: 491.54
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 7672022
MDL No.: MFCD00151937
Formula:
C₃₁H₂₅NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00151937 CAS Number: 117666-96-3 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...