tert-Butyl (2-hydroxyethyl)(methyl)carbamate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(tert-Butoxycarbonyl)-N-methylethanolamine, N-Boc-N-methyl-2-aminoethanol, tert-Butyl (2-Hydroxyethyl)(methyl)carbamate, N-(tert-Butoxycarbonyl)-N-methyl-2-aminoethanol, N-Boc-N-methylethanolamine, N-Boc-N-methylglycinol, N-(tert-Butoxycarbonyl)-N-methylglycinol, (2-Hydroxyethyl)(methyl)carbamic acid tert-Butyl ester

H66660-03 H66660-06 H66660-14
AAH66660-03EA 33.9 USD
AAH66660-03 AAH66660-06 AAH66660-14
tert-Butyl (2-hydroxyethyl)(methyl)carbamate 95%
tert-Butyl (2-hydroxyethyl)(methyl)carbamate

N-Boc-N-methylethanolamine, 95%


Purity: 95% CAS No.: 57561-39-4 Molecular Formula: C8H17NO3 Formula Weight: 175.23 Storage Temperature: -30°C to -10°C Physical Form: Liquid Appearance: Colorless to pale yellow Beilstein Reference: 3537964 MDL No.: MFCD03425857

Formula: C8H17NO3
MW: 175.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03425857
CAS Number: 57561-39-4

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR