N-Fmoc-D-glutamic acid 5-tert-butyl ester 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate, 5-tert-Butyl N-Fmoc-D-glutamate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid 5-tert-Butyl Ester, Fmoc-D-Glu(OtBu)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid
N-Fmoc-D-glutamic acid 5-tert-butyl ester, 98%
Purity: 98%
CAS No.: 104091-08-9
Molecular Formula: C24H27NO6
Formula Weight: 425.48
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 83-89°
Optical Rotation: +4°(c=1 in acetic acid soln.)
MDL No.: MFCD00077055
Formula:
C₂₄H₂₇NO₆ Melting Pt: 83…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077055 CAS Number: 104091-08-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...