D-Valine ethyl ester hydrochloride 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-D-Val-OEt·HCl, (R)-Ethyl 2-amino-3-methylbutanoate hydrochloride
H66619-03
H66619-06
H66619-14
AAH66619-03EA
36.4
USD
AAH66619-03
AAH66619-06
AAH66619-14
D-Valine ethyl ester hydrochloride 97%
D-Valine ethyl ester hydrochloride
D-Valine ethyl ester hydrochloride, 97%
Purity: 97%
CAS No.: 73913-64-1
Molecular Formula: C7H15NO2.HCl
Formula Weight: 181.66
Physical Form: Solid
Melting Point: 102-104°
Beilstein Reference: 3912412
MDL No.: MFCD00191043
Formula:
C₇H₁₆ClNO₂ Melting Pt: 102…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191043 CAS Number: 73913-64-1 EINECS: 277-635-7 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...