Fmoc-allo-Thr(tBu)-OH 97%
Supplier: Thermo Scientific Chemicals
N-Fmoc-O-tert-butyl-L-allo-threonine, 97%
Purity: 97%
CAS No.: 201481-37-0
Molecular Formula: C23H27NO5
Formula Weight: 397.47
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD00237371
Formula:
C₂₃H₂₇NO₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD00237371 CAS Number: 201481-37-0 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...