Fmoc-Asp-OBzl 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-L-aspartic acid 1-benzyl ester, Fmoc-Asp-Obzl

H66558-03 H66558-06
AAH66558-03EA 48.3 USD
AAH66558-03 AAH66558-06 AAH66558-MD
Fmoc-Asp-OBzl 95%
Fmoc-Asp-OBzl

N-Fmoc-L-aspartic acid 1-benzyl ester, 95%


Purity: 95% CAS No.: 86060-83-5 Molecular Formula: C26H23NO6 Formula Weight: 445.47 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Beilstein Reference: 4606552 MDL No.: MFCD00198201

Formula: C₂₆H₂₃NO₆
MW: 445.471 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00198201
CAS Number: 86060-83-5

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR