2-(Iodomethyl)Oxetane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Iodomethyl)oxetane
H66525-03
H66525-MD
AAH66525-03EA
1154
USD
AAH66525-03
AAH66525-MD
2-(Iodomethyl)Oxetane 97%
2-(Iodomethyl)Oxetane
2-(Iodomethyl)oxetane, 97%
Purity: 97%
CAS No.: 121138-00-9
Molecular Formula: C4H7IO
Formula Weight: 198.00
Physical Form: Liquid
Sensitivity: Light Sensitive
Appearance: Colorless to pale yellow
MDL No.: MFCD11226849
Formula:
C₄H₇IO MW: 198.003 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11226849 CAS Number: 121138-00-9 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...